siRNA Details
| Gene Symbol: Ptrh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013860.1 | NP_001013882.1 | 2 | 505 | ||
| GenBank mRNA | BC083801.1 | - | 2 | 505 | 7108916 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptrh2| Gene Symbol: Ptrh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013860.1 | NP_001013882.1 | 2 | 505 | ||
| GenBank mRNA | BC083801.1 | - | 2 | 505 | 7108916 | |