siRNA Details
| Gene Symbol: Dhx40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005873.1 | NP_001005873.1 | 9 | 1236 | ||
| GenBank mRNA | BC083821.1 | - | 9 | 1236 | 7111216 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhx40| Gene Symbol: Dhx40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005873.1 | NP_001005873.1 | 9 | 1236 | ||
| GenBank mRNA | BC083821.1 | - | 9 | 1236 | 7111216 | |