siRNA Details
| Gene Symbol: Kif2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005874.2 | NP_001005874.2 | 1 | 337 | ||
| GenBank mRNA | BC083841.1 | - | 1 | 334 | 7114321 | |
| CK594948.1 | - | 1 | 356 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif2b| Gene Symbol: Kif2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005874.2 | NP_001005874.2 | 1 | 337 | ||
| GenBank mRNA | BC083841.1 | - | 1 | 334 | 7114321 | |
| CK594948.1 | - | 1 | 356 | |||