siRNA Details
| Gene Symbol: Samd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024966.1 | NP_001020137.1 | 10 | 1240 | ||
| GenBank mRNA | BC091329.1 | - | 10 | 1240 | 7313977 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Samd14| Gene Symbol: Samd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024966.1 | NP_001020137.1 | 10 | 1240 | ||
| GenBank mRNA | BC091329.1 | - | 10 | 1240 | 7313977 | |