siRNA Details
| Gene Symbol: Spop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100496.1 | NP_001093966.1 | 6 | 683 | ||
| GenBank mRNA | BC166743.1 | - | 6 | 683 | 7938440 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spop| Gene Symbol: Spop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100496.1 | NP_001093966.1 | 6 | 683 | ||
| GenBank mRNA | BC166743.1 | - | 6 | 683 | 7938440 | |