siRNA Details
| Gene Symbol: Snf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007804.1 | NP_001007805.1 | 8 | 751 | ||
| XM_006247185.1 | XP_006247247.1 | 8 | 884 | |||
| GenBank mRNA | BC086364.1 | - | 8 | 794 | 7304427 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snf8| Gene Symbol: Snf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007804.1 | NP_001007805.1 | 8 | 751 | ||
| XM_006247185.1 | XP_006247247.1 | 8 | 884 | |||
| GenBank mRNA | BC086364.1 | - | 8 | 794 | 7304427 | |