siRNA Details
| Gene Symbol: Ccr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199489.4 | NP_955783.1 | 3 | 981 | ||
| GenBank mRNA | AY379972.1 | - | 3 | 984 | ||
| BC089762.1 | - | 968 | 7381637 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccr7| Gene Symbol: Ccr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199489.4 | NP_955783.1 | 3 | 981 | ||
| GenBank mRNA | AY379972.1 | - | 3 | 984 | ||
| BC089762.1 | - | 968 | 7381637 | |||