siRNA Details
| Gene Symbol: Krt35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008820.1 | NP_001008820.1 | 7 | 1253 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt35| Gene Symbol: Krt35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008820.1 | NP_001008820.1 | 7 | 1253 | ||