siRNA Details
| Gene Symbol: Dhx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047844.1 | NP_001041309.1 | 15 | 2533 | ||
| XM_006247286.2 | XP_006247348.1 | 17 | 2913 | |||
| GenBank mRNA | AY383690.1 | - | 15 | 2533 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhx8| Gene Symbol: Dhx8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047844.1 | NP_001041309.1 | 15 | 2533 | ||
| XM_006247286.2 | XP_006247348.1 | 17 | 2913 | |||
| GenBank mRNA | AY383690.1 | - | 15 | 2533 | ||