siRNA Details
| Gene Symbol: Pyy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034080.1 | NP_001029252.1 | 3, 4 | 339 | ||
| GenBank mRNA | AA998735.1 | - | 1, 2 | 193 | ||
| AB238226.1 | - | 3, 4 | 339 | |||
| M17523.1 | - | 2, 3 | 318 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pyy| Gene Symbol: Pyy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034080.1 | NP_001029252.1 | 3, 4 | 339 | ||
| GenBank mRNA | AA998735.1 | - | 1, 2 | 193 | ||
| AB238226.1 | - | 3, 4 | 339 | |||
| M17523.1 | - | 2, 3 | 318 | |||