siRNA Details
| Gene Symbol: Prp2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768296.1 | XP_008766518.1 | 11 | 14193 | ||
| GenBank mRNA | BC101875.1 | - | 8 | 1807 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prp2l1| Gene Symbol: Prp2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768296.1 | XP_008766518.1 | 11 | 14193 | ||
| GenBank mRNA | BC101875.1 | - | 8 | 1807 | ||