siRNA Details
Gene Symbol: Lyzl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135833.1 | NP_001129305.2 | 2 | 120 | ||
XM_006247503.2 | XP_006247565.1 | 2 | 1633 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lyzl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135833.1 | NP_001129305.2 | 2 | 120 | ||
XM_006247503.2 | XP_006247565.1 | 2 | 1633 |