siRNA Details
| Gene Symbol: Cep112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105849.1 | NP_001099319.1 | 9 | 973 | ||
| XM_008768317.1 | XP_008766539.1 | 9 | 1007 | |||
| XM_008768318.1 | XP_008766540.1 | 9 | 1145 | |||
| XM_008768319.1 | XP_008766541.1 | 9 | 1007 | |||
| XM_008768320.1 | XP_008766542.1 | 9 | 1087 | |||
| XM_008768321.1 | XP_008766543.1 | 9 | 1581 | |||

Cep112
Genomic Map