siRNA Details
| Gene Symbol: Hn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005876.1 | NP_001005876.1 | 1 | 123 | ||
| GenBank mRNA | BC079311.1 | - | 1 | 123 | 7134794 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hn1| Gene Symbol: Hn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005876.1 | NP_001005876.1 | 1 | 123 | ||
| GenBank mRNA | BC079311.1 | - | 1 | 123 | 7134794 | |