siRNA Details
| Gene Symbol: RGD1308065 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077666.1 | NP_001071134.1 | 5 | 701 | ||
| XM_008768856.1 | XP_008767078.1 | 5 | 778 | |||
| XM_008768858.1 | XP_008767080.1 | 5 | 790 | |||
| GenBank mRNA | BC127537.1 | - | 5 | 701 | 7135417 | |

RGD1308065
Genomic Map