siRNA Details
| Gene Symbol: Ccdc116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024740.1 | NP_001019911.1 | 3 | 384 | ||
| XM_006248676.2 | XP_006248738.1 | 3 | 506 | |||
| GenBank mRNA | BC097459.1 | - | 3 | 384 | 7457223 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc116| Gene Symbol: Ccdc116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024740.1 | NP_001019911.1 | 3 | 384 | ||
| XM_006248676.2 | XP_006248738.1 | 3 | 506 | |||
| GenBank mRNA | BC097459.1 | - | 3 | 384 | 7457223 | |