siRNA Details
| Gene Symbol: Parl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035249.1 | NP_001030326.1 | 1, 2 | 165 | ||
| GenBank mRNA | BC088226.1 | - | 1, 2 | 122 | ||
| BC105907.1 | - | 1, 2 | 165 | |||
| BC126098.1 | - | 1, 2 | 141 | |||
| BC158564.1 | - | 1, 2 | 149 | 8374563 | ||
| FQ214115.1 | - | 1, 2 | 160 | |||

Parl
Genomic Map