siRNA Details
| Gene Symbol: Vps8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248575.2 | XP_006248637.1 | 48 | 4427 | ||
| XM_006248577.2 | XP_006248639.1 | 48 | 4419 | |||
| XM_006248578.2 | XP_006248640.1 | 48 | 4451 | |||
| XM_006248579.2 | XP_006248641.1 | 47 | 4421 | |||
| XM_008768830.1 | XP_008767052.1 | 48 | 4626 | |||

Vps8
Genomic Map