siRNA Details
| Gene Symbol: LOC287992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY383659.1 | - | 1357 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC287992| Gene Symbol: LOC287992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY383659.1 | - | 1357 | |||