siRNA Details
| Gene Symbol: Tmem44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768813.1 | XP_008767035.1 | 8 | 2127 | ||
| XM_213600.8 | XP_213600.7 | 8 | 2127 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem44| Gene Symbol: Tmem44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768813.1 | XP_008767035.1 | 8 | 2127 | ||
| XM_213600.8 | XP_213600.7 | 8 | 2127 | |||