siRNA Details
| Gene Symbol: Fam43a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039002.1 | NP_001034091.1 | 1 | 676 | ||
| GenBank mRNA | BC099231.1 | - | 1 | 676 | 7442928 | |
| Gene Symbol: LOC100909474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752202.3 | XP_003752250.1 | 1 | 501 | ||

Fam43a
Genomic Map