siRNA Details
Gene Symbol: Poglut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100652.1 | NP_001094122.1 | 8 | 864 | ||
GenBank mRNA | BC091408.1 | - | 7 | 636 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Poglut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100652.1 | NP_001094122.1 | 8 | 864 | ||
GenBank mRNA | BC091408.1 | - | 7 | 636 |