siRNA Details
| Gene Symbol: Retnlg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181625.1 | NP_853656.1 | 1 | 68 | ||
| GenBank mRNA | AJ514932.1 | - | 1 | 68 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Retnlg| Gene Symbol: Retnlg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181625.1 | NP_853656.1 | 1 | 68 | ||
| GenBank mRNA | AJ514932.1 | - | 1 | 68 | ||