siRNA Details
| Gene Symbol: Pigp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099758.1 | NP_001093228.1 | 3 | 231 | ||
| XM_006248042.2 | XP_006248104.1 | 2 | 302 | |||
| XM_006248043.2 | XP_006248105.1 | 3 | 321 | |||
| GenBank mRNA | BC166404.1 | - | 3 | 321 | 9099247 | |
| BF555819.1 | - | 3 | 238 | |||
| CF978233.1 | - | 3 | 196 | |||

Pigp
Genomic Map