siRNA Details
| Gene Symbol: Ripply3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105892.1 | NP_001099362.1 | 3 | 297 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ripply3| Gene Symbol: Ripply3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105892.1 | NP_001099362.1 | 3 | 297 | ||