siRNA Details
| Gene Symbol: Cryzl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013044.1 | NP_001013062.1 | 3 | 130 | ||
| XM_006248045.2 | XP_006248107.1 | 3 | 746 | |||
| XM_006248046.2 | XP_006248108.1 | 1 | 29 | |||
| GenBank mRNA | BC083853.1 | - | 3 | 130 | 7124012 | |

Cryzl1
Genomic Map