siRNA Details
| Gene Symbol: Tmprss15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105895.1 | NP_001099365.1 | 16, 17 | 1844 | ||
| XM_008768527.1 | XP_008766749.1 | 14, 15 | 1658 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss15| Gene Symbol: Tmprss15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105895.1 | NP_001099365.1 | 16, 17 | 1844 | ||
| XM_008768527.1 | XP_008766749.1 | 14, 15 | 1658 | |||