siRNA Details
| Gene Symbol: Krtap15-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105896.1 | NP_001099366.1 | 1 | 110 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap15-1| Gene Symbol: Krtap15-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105896.1 | NP_001099366.1 | 1 | 110 | ||