siRNA Details
| Gene Symbol: Pomp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100942.1 | NP_001094412.1 | 6 | 413 | ||
| GenBank mRNA | BC161800.1 | - | 6 | 456 | 9026464 | |
| EF535528.1 | - | 6 | 413 | |||
| FQ217734.1 | - | 5 | 404 | |||
| FQ224264.1 | - | 6 | 450 | |||
| Gene Symbol: LOC100911238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248840.1 | XP_006248902.1 | 6 | 500 | ||

Pomp
Genomic Map