siRNA Details
Gene Symbol: Nptx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034199.1 | NP_001029371.1 | 3 | 939 | ||
GenBank mRNA | CO395871.1 | - | 2 | 267 | ||
S82649.1 | - | 873 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nptx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034199.1 | NP_001029371.1 | 3 | 939 | ||
GenBank mRNA | CO395871.1 | - | 2 | 267 | ||
S82649.1 | - | 873 |