siRNA Details
| Gene Symbol: Eif3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031640.1 | NP_001026810.1 | 12 | 1703 | ||
| GenBank mRNA | BC098728.1 | - | 12 | 1703 | 7381504 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif3b| Gene Symbol: Eif3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031640.1 | NP_001026810.1 | 12 | 1703 | ||
| GenBank mRNA | BC098728.1 | - | 12 | 1703 | 7381504 | |