siRNA Details
| Gene Symbol: Akap17a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127246.1 | NP_001120718.1 | 5 | 1870 | ||
| XM_006248998.2 | XP_006249060.1 | 5 | 2583 | |||
| XM_006248999.2 | XP_006249061.1 | 5 | 2427 | |||
| XM_006249000.2 | XP_006249062.1 | 5 | 2518 | |||
| GenBank mRNA | BC166722.1 | - | 5 | 1870 | 7933458 | |

Akap17a
Genomic Map