siRNA Details
| Gene Symbol: RGD1305455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024969.2 | NP_001020140.2 | 5 | 954 | ||
| GenBank mRNA | BC091263.1 | - | 4 | 824 | ||
| DY317491.1 | - | 322 | ||||
| FQ211718.1 | - | 5 | 1000 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1305455| Gene Symbol: RGD1305455 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024969.2 | NP_001020140.2 | 5 | 954 | ||
| GenBank mRNA | BC091263.1 | - | 4 | 824 | ||
| DY317491.1 | - | 322 | ||||
| FQ211718.1 | - | 5 | 1000 | |||