siRNA Details
| Gene Symbol: Fis1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105919.1 | NP_001099389.1 | 4 | 596 | ||
| XM_006249121.2 | XP_006249183.1 | 4 | 662 | |||
| XM_006249122.2 | XP_006249184.1 | 4 | 616 | |||
| XM_006249123.2 | XP_006249185.1 | 4 | 642 | |||
| GenBank mRNA | BC167057.1 | - | 4 | 609 | 9098277 | |
| FQ213833.1 | - | 4 | 422 | |||
| FQ222246.1 | - | 4 | 408 | |||
| FQ224250.1 | - | 4 | 419 | |||

Fis1
Genomic Map