siRNA Details
| Gene Symbol: Ccl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013045.1 | NP_001013063.1 | 3 | 359 | ||
| GenBank mRNA | BC087576.1 | - | 3 | 359 | 7307568 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccl24| Gene Symbol: Ccl24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013045.1 | NP_001013063.1 | 3 | 359 | ||
| GenBank mRNA | BC087576.1 | - | 3 | 359 | 7307568 | |