siRNA Details
| Gene Symbol: Eif4h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006957.2 | NP_001006958.1 | 2, 3 | 281 | ||
| XM_006249134.1 | XP_006249196.1 | 2, 3 | 504 | |||
| GenBank mRNA | BC083818.1 | - | 2, 3 | 273 | 7110979 | |
| CB718613.1 | - | 2, 3 | 281 | |||

Eif4h
Genomic Map