siRNA Details
| Gene Symbol: Wbscr28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249213.2 | XP_006249275.1 | 2 | 1085 | ||
| XM_213758.8 | XP_213758.4 | 2 | 444 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wbscr28| Gene Symbol: Wbscr28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006249213.2 | XP_006249275.1 | 2 | 1085 | ||
| XM_213758.8 | XP_213758.4 | 2 | 444 | |||