siRNA Details
| Gene Symbol: Cct6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033684.1 | NP_001028856.1 | 11 | 1340 | ||
| GenBank mRNA | BC091282.1 | - | 5 | 462 | ||
| BC104703.1 | - | 11 | 1340 | 7300674 | ||
| FQ216833.1 | - | 11 | 1358 | |||
| FQ218432.1 | - | 11 | 1387 | |||

Cct6a
Genomic Map