siRNA Details
| Gene Symbol: Arl6ip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025630.1 | NP_001020801.1 | 3 | 858 | ||
| GenBank mRNA | BC097374.1 | - | 3 | 858 | 7444619 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arl6ip4| Gene Symbol: Arl6ip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025630.1 | NP_001020801.1 | 3 | 858 | ||
| GenBank mRNA | BC097374.1 | - | 3 | 858 | 7444619 | |