siRNA Details
| Gene Symbol: Rab35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013046.1 | NP_001013064.1 | 2 | 177 | ||
| XM_008769221.1 | XP_008767443.1 | 2 | 227 | |||
| GenBank mRNA | BC085769.1 | - | 2 | 177 | 7135343 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab35| Gene Symbol: Rab35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013046.1 | NP_001013064.1 | 2 | 177 | ||
| XM_008769221.1 | XP_008767443.1 | 2 | 227 | |||
| GenBank mRNA | BC085769.1 | - | 2 | 177 | 7135343 | |