siRNA Details
| Gene Symbol: Unc119b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105934.1 | NP_001099404.1 | 4 | 602 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc119b| Gene Symbol: Unc119b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105934.1 | NP_001099404.1 | 4 | 602 | ||