siRNA Details
| Gene Symbol: Gltp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134413.1 | NP_001127885.1 | 2 | 204 | ||
| GenBank mRNA | BC158884.1 | - | 2 | 204 | 7938728 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gltp| Gene Symbol: Gltp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134413.1 | NP_001127885.1 | 2 | 204 | ||
| GenBank mRNA | BC158884.1 | - | 2 | 204 | 7938728 | |