siRNA Details
| Gene Symbol: Pgam5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025272.1 | NP_001020443.1 | 3, 4 | 527 | ||
| XM_006249534.2 | XP_006249596.1 | 3, 4 | 559 | |||
| GenBank mRNA | BC092607.1 | - | 3, 4 | 527 | 7309810 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgam5| Gene Symbol: Pgam5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025272.1 | NP_001020443.1 | 3, 4 | 527 | ||
| XM_006249534.2 | XP_006249596.1 | 3, 4 | 559 | |||
| GenBank mRNA | BC092607.1 | - | 3, 4 | 527 | 7309810 | |