siRNA Details
Gene Symbol: Sgsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105937.2 | NP_001099407.2 | 11 | 1098 | ||
XM_008769322.1 | XP_008767544.1 | 11 | 1437 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sgsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105937.2 | NP_001099407.2 | 11 | 1098 | ||
XM_008769322.1 | XP_008767544.1 | 11 | 1437 |