siRNA Details
| Gene Symbol: Spryd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037765.1 | NP_001032854.1 | 2 | 258 | ||
| XM_006240746.1 | XP_006240808.1 | 3 | 434 | |||
| GenBank mRNA | BC099138.1 | - | 2 | 258 | 7456187 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spryd4| Gene Symbol: Spryd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037765.1 | NP_001032854.1 | 2 | 258 | ||
| XM_006240746.1 | XP_006240808.1 | 3 | 434 | |||
| GenBank mRNA | BC099138.1 | - | 2 | 258 | 7456187 | |