siRNA Details
Gene Symbol: Rab5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079936.1 | NP_001073405.1 | 4 | 608 | ||
XM_006240750.2 | XP_006240812.1 | 4 | 531 | |||
XM_006240751.2 | XP_006240813.1 | 5 | 629 | |||
XM_006240752.2 | XP_006240814.1 | 4 | 756 | |||
GenBank mRNA | BC129101.1 | - | 4 | 608 | 7098463 | |
FQ211489.1 | - | 4 | 746 |
