siRNA Details
Gene Symbol: Gadd45gip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100504.1 | NP_001093974.1 | 2 | 661 | ||
GenBank mRNA | BC083178.1 | - | 2 | 653 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gadd45gip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100504.1 | NP_001093974.1 | 2 | 661 | ||
GenBank mRNA | BC083178.1 | - | 2 | 653 |