siRNA Details
Gene Symbol: RGD1564093 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105946.1 | NP_001099416.1 | 1 | 360 | ||
GenBank mRNA | FQ231178.1 | - | 1 | 299 | ||
FQ235293.1 | - | 1 | 299 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1564093 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105946.1 | NP_001099416.1 | 1 | 360 | ||
GenBank mRNA | FQ231178.1 | - | 1 | 299 | ||
FQ235293.1 | - | 1 | 299 |