siRNA Details
| Gene Symbol: Cdk18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100506.1 | NP_001093976.1 | 11, 12 | 1117 | ||
| XM_006249750.2 | XP_006249812.1 | 12, 13 | 1204 | |||
| XM_006249751.2 | XP_006249813.1 | 12, 13 | 1265 | |||
| XM_006249752.2 | XP_006249814.1 | 12, 13 | 1725 | |||
| XM_006249753.2 | XP_006249815.1 | 12, 13 | 1238 | |||
| XM_008769441.1 | XP_008767663.1 | 12, 13 | 1543 | |||
| GenBank mRNA | AB005541.1 | - | 12, 13 | 1197 | ||
| BC082045.1 | - | 11, 12 | 1117 | 7131722 | ||

Cdk18
Genomic Map