siRNA Details
Gene Symbol: RGD1308112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006249895.2 | XP_006249957.1 | 5 | 1685 | ||
XM_008769575.1 | XP_008767797.1 | 4 | 1602 | |||
XM_008769576.1 | XP_008767798.1 | 5 | 1680 |
